siRNA Details
| Gene Symbol: LOC105372751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135520.1 | 6 | 2291 | |||
| GenBank mRNA | AK124194.1 | - | 6 | 2291 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC024236.1 | - | 1 | 163 | Chr. 17 - 62047228 - 62048427 [+] on Build GRCh38 | |

LOC105372751
Genomic Map