siRNA Details
| Gene Symbol: LOC153811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037953.1 | - | 1 | 1077 | ||
| Gene Symbol: RNF130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534593.2 | XP_011532895.1 | 8 | 2841 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM701594.1 | - | 1 | 146 | Chr. 5 - 179918911 - 179919309 [-] on Build GRCh38 | |

LOC153811
Genomic Map