siRNA Details
| Gene Symbol: LOC153811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037953.1 | - | 1 | 839 | ||
| Gene Symbol: RNF130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534593.2 | XP_011532895.1 | 8 | 2603 | ||

LOC153811
Genomic Map