siRNA Details
Gene Symbol: LOC283332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC121095.1 | - | 1 | 140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB039238.1 | - | 2 | 189 | Chr. 12 - 49910760 - 49911858 [-] on Build GRCh38 |
