siRNA Details
| Gene Symbol: ZNF641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005268637.3 | XP_005268694.1 | 5 | 5978 | ||
| XM_005268638.3 | XP_005268695.1 | 6 | 6218 | |||
| XM_005268639.2 | XP_005268696.1 | 5 | 5869 | |||
| XM_011537897.1 | XP_011536199.1 | 6 | 6198 | |||
| XM_011537898.1 | XP_011536200.1 | 6 | 6126 | |||
| XM_017018797.1 | XP_016874286.1 | 5 | 6022 | |||
| XM_017018798.1 | XP_016874287.1 | 6 | 6052 | |||
| GenBank mRNA | AK054984.1 | - | 1 | 883 | ||
| AK127710.1 | - | 1 | 1830 | |||

ZNF641
Genomic Map