siRNA Details
| Gene Symbol: LOC102724765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC070382.1 | - | 1 | 268 | ||
| Gene Symbol: FAM65B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286446.1 | NP_001273375.1 | 1 | 266 | ||
| XM_006715275.2 | XP_006715338.1 | 1 | 380 | |||
| XM_006715281.3 | XP_006715344.1 | 1 | 380 | |||
| XM_011515007.1 | XP_011513309.1 | 1 | 386 | |||
| XM_011515012.1 | XP_011513314.1 | 1 | 380 | |||
| GenBank mRNA | AK300950.1 | - | 1 | 116 | ||
| DC406827.1 | - | 1 | 267 | |||

LOC102724765
Genomic Map