siRNA Details
| Gene Symbol: FAM78B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB593134.1 | - | 2 | 2778 | ||
| AK055347.1 | - | 1 | 817 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK430134.1 | - | 1 | 464 | Chr. 1 - 166058484 - 166059119 [-] on Build GRCh38 | |

FAM78B
Genomic Map