siRNA Details
| Gene Symbol: FAM78B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320302.1 | NP_001307231.1 | 2 | 2757 | ||
| NR_135199.1 | 3 | 3689 | ||||
| XM_017000417.1 | XP_016855906.1 | 3 | 7895 | |||
| XM_017000418.1 | XP_016855907.1 | 3 | 3237 | |||
| GenBank mRNA | AB593134.1 | - | 2 | 2757 | ||
| AK055347.1 | - | 1 | 796 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK430134.1 | - | 1 | 443 | Chr. 1 - 166058484 - 166059119 [-] on Build GRCh38 | |

FAM78B
Genomic Map