siRNA Details
| Gene Symbol: DLEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AJ412029.1 | - | LIT1923 BCMS |
6 | 1016 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AJ412022.3 | - | 1 | 509 | Chr. 13 - 50723522 - 50724063 [+] on Build GRCh38 | |

DLEU1
Genomic Map