siRNA Details
| Gene Symbol: ZNF346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308213.1 | NP_001295142.1 | 9 | 2474 | ||
| NM_001308219.1 | NP_001295148.1 | 5 | 1974 | |||
| NM_001308223.1 | NP_001295152.1 | 4 | 1871 | |||
| XM_005265860.2 | XP_005265917.1 | 9 | 2467 | |||
| XM_011534494.1 | XP_011532796.1 | 8 | 2373 | |||
| XM_017009294.1 | XP_016864783.1 | 8 | 2447 | |||
| XM_017009295.1 | XP_016864784.1 | 8 | 2399 | |||
| GenBank mRNA | AK022292.1 | - | 1 | 2158 | ||
| AK131469.1 | - | 10 | 2419 | |||
| CR627467.1 | - | 1 | 1324 | |||

ZNF346
Genomic Map