siRNA Details
| Gene Symbol: LOC157860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK025743.1 | - | 1 | 2166 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX103959.1 | - | 1 | 484 | Chr. 8 - 37597497 - 37598169 [-] on Build GRCh38 | |

LOC157860
Genomic Map