siRNA Details
| Gene Symbol: FAM63B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040450.2 | NP_001035540.1 | 9 | 6655 | ||
| NM_001040453.2 | NP_001035543.1 | 9 | 6482 | |||
| XM_011521687.1 | XP_011519989.1 | 10 | 6707 | |||
| XM_011521688.1 | XP_011519990.1 | 10 | 6704 | |||
| XM_011521689.1 | XP_011519991.1 | 10 | 6605 | |||
| XM_011521690.1 | XP_011519992.1 | 9 | 6551 | |||
| XM_017022332.1 | XP_016877821.1 | 8 | 6461 | |||
| XM_017022333.1 | XP_016877822.1 | 7 | 6407 | |||
| XM_017022334.1 | XP_016877823.1 | 7 | 6404 | |||
| GenBank mRNA | AK021710.1 | - | 1 | 725 | ||

FAM63B
Genomic Map