siRNA Details
| Gene Symbol: LOC101928911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY927642.1 | - | 3 | 1045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928911| Gene Symbol: LOC101928911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY927642.1 | - | 3 | 1045 | ||