siRNA Details
| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007202.3 | NP_009133.2 | 15 | 3515 | ||
| XM_006721432.3 | XP_006721495.1 | 14 | 3336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AKAP10| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007202.3 | NP_009133.2 | 15 | 3515 | ||
| XM_006721432.3 | XP_006721495.1 | 14 | 3336 | |||