siRNA Details
| Gene Symbol: MORF4L2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038978.1 | 2 | 469 | |||
| GenBank mRNA | AK124135.1 | - | 1 | 1392 | ||
| BI832939.1 | - | 2 | 469 | |||
| DA678830.1 | - | 2 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MORF4L2-AS1| Gene Symbol: MORF4L2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038978.1 | 2 | 469 | |||
| GenBank mRNA | AK124135.1 | - | 1 | 1392 | ||
| BI832939.1 | - | 2 | 469 | |||
| DA678830.1 | - | 2 | 439 | |||