siRNA Details
| Gene Symbol: ZNF528-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022990.1 | - | 1 | 1573 | ||
| AK097590.1 | - | 5 | 1089 | |||
| BX537909.1 | - | 4 | 5028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF528-AS1| Gene Symbol: ZNF528-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022990.1 | - | 1 | 1573 | ||
| AK097590.1 | - | 5 | 1089 | |||
| BX537909.1 | - | 4 | 5028 | |||