siRNA Details
| Gene Symbol: CDS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003818.3 | NP_003809.1 | 13 | 8082 | ||
| XM_006723660.1 | XP_006723723.1 | 13 | 8093 | |||
| XM_011529390.1 | XP_011527692.1 | 14 | 8405 | |||
| GenBank mRNA | AK022998.1 | - | 1 | 1426 | ||
| AK123831.1 | - | 1 | 2378 | |||
| BC025346.1 | - | 1 | 15 | |||

CDS2
Genomic Map