siRNA Details
| Gene Symbol: GALNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003237.1 | XP_016858726.1 | 10 | 4685 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK022198.1 | - | 1 | 321 | Chr. 2 - 157312307 - 157314211 [+] on Build GRCh38 | |

GALNT5
Genomic Map