siRNA Details
| Gene Symbol: TACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY139007.1 | - | 1 | 439 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK129750.1 | - | 1 | 995 | Chr. 8 - 38838727 - 38840059 [+] on Build GRCh38 | |

TACC1
Genomic Map