siRNA Details
| Gene Symbol: FAM98B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173611.3 | NP_775882.2 | 8 | 2789 | ||
| GenBank mRNA | BC045690.1 | - | 1 | 1477 | ||
| DA118586.1 | - | 1 | 212 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR607099.1 | - | 1 | 704 | Chr. 15 - 38485408 - 38487080 [+] on Build GRCh38 | |

FAM98B
Genomic Map