siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 86 | 20718 | ||
XM_006721206.2 | XP_006721269.1 | 86 | 20612 | |||
XM_011523146.2 | XP_011521448.1 | 87 | 20747 | |||
XM_011523147.1 | XP_011521449.1 | 87 | 20714 | |||
XM_011523148.1 | XP_011521450.1 | 87 | 20787 | |||
XM_011523151.1 | XP_011521453.1 | 86 | 20644 | |||
XM_011523152.1 | XP_011521454.1 | 49 | 14471 | |||
XM_011523155.2 | XP_011521457.1 | 42 | 13472 | |||
XM_017023346.1 | XP_016878835.1 | 86 | 20890 | |||
XM_017023347.1 | XP_016878836.1 | 74 | 19129 | |||
XM_017023348.1 | XP_016878837.1 | 74 | 18869 | |||
GenBank mRNA | AK125886.1 | - | 2 | 3454 |
