siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 86 | 20998 | ||
XM_006721206.2 | XP_006721269.1 | 86 | 20892 | |||
XM_011523146.2 | XP_011521448.1 | 87 | 21027 | |||
XM_011523147.1 | XP_011521449.1 | 87 | 20994 | |||
XM_011523148.1 | XP_011521450.1 | 87 | 21067 | |||
XM_011523151.1 | XP_011521453.1 | 86 | 20924 | |||
XM_011523152.1 | XP_011521454.1 | 49 | 14751 | |||
XM_011523155.2 | XP_011521457.1 | 42 | 13752 | |||
XM_017023346.1 | XP_016878835.1 | 86 | 21170 | |||
XM_017023347.1 | XP_016878836.1 | 74 | 19409 | |||
XM_017023348.1 | XP_016878837.1 | 74 | 19149 | |||
GenBank mRNA | AK125886.1 | - | 2 | 3734 |
