siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 86 | 17808 | ||
XM_006721206.2 | XP_006721269.1 | 86 | 17702 | |||
XM_011523146.2 | XP_011521448.1 | 87 | 17837 | |||
XM_011523147.1 | XP_011521449.1 | 87 | 17804 | |||
XM_011523148.1 | XP_011521450.1 | 87 | 17877 | |||
XM_011523151.1 | XP_011521453.1 | 86 | 17734 | |||
XM_011523152.1 | XP_011521454.1 | 49 | 11561 | |||
XM_011523155.2 | XP_011521457.1 | 42 | 10562 | |||
XM_017023346.1 | XP_016878835.1 | 86 | 17980 | |||
XM_017023347.1 | XP_016878836.1 | 74 | 16219 | |||
XM_017023348.1 | XP_016878837.1 | 74 | 15959 | |||
GenBank mRNA | AK125886.1 | - | 1 | 815 |
