siRNA Details
| Gene Symbol: HYDIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270974.2 | NP_001257903.1 | 86 | 19075 | ||
| XM_006721206.2 | XP_006721269.1 | 86 | 18969 | |||
| XM_011523146.2 | XP_011521448.1 | 87 | 19104 | |||
| XM_011523147.1 | XP_011521449.1 | 87 | 19071 | |||
| XM_011523148.1 | XP_011521450.1 | 87 | 19144 | |||
| XM_011523151.1 | XP_011521453.1 | 86 | 19001 | |||
| XM_011523152.1 | XP_011521454.1 | 49 | 12828 | |||
| XM_011523155.2 | XP_011521457.1 | 42 | 11829 | |||
| XM_017023346.1 | XP_016878835.1 | 86 | 19247 | |||
| XM_017023347.1 | XP_016878836.1 | 74 | 17486 | |||
| XM_017023348.1 | XP_016878837.1 | 74 | 17226 | |||
| GenBank mRNA | AK125886.1 | - | 2 | 1811 | ||

HYDIN
Genomic Map