siRNA Details
| Gene Symbol: LOC101928751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125964.1 | 2 | 720 | |||
| GenBank mRNA | BC031108.1 | - | 2 | 720 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928751| Gene Symbol: LOC101928751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125964.1 | 2 | 720 | |||
| GenBank mRNA | BC031108.1 | - | 2 | 720 | ||