siRNA Details
Gene Symbol: BRAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012558.1 | XP_016868047.1 | 10 | 1272 | ||
XM_017012559.1 | XP_016868048.1 | 10 | 1272 | |||
GenBank mRNA | AJ276310.1 | - | 1 | 91 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
