siRNA Details
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012558.1 | XP_016868047.1 | 10, 11 | 1298 | ||
| XM_017012559.1 | XP_016868048.1 | 10, 11 | 1298 | |||
| GenBank mRNA | AJ276310.1 | - | 117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRAF| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012558.1 | XP_016868047.1 | 10, 11 | 1298 | ||
| XM_017012559.1 | XP_016868048.1 | 10, 11 | 1298 | |||
| GenBank mRNA | AJ276310.1 | - | 117 | |||