siRNA Details
| Gene Symbol: PLIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014259.1 | XP_016869748.1 | 8 | 1403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLIN2| Gene Symbol: PLIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014259.1 | XP_016869748.1 | 8 | 1403 | ||