siRNA Details
Gene Symbol: THUMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128398.1 | - | 1 | 2431 | ||
BC036698.1 | - | 1 | 2023 | |||
BC052961.1 | - | 1 | 2025 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: THUMPD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128398.1 | - | 1 | 2431 | ||
BC036698.1 | - | 1 | 2023 | |||
BC052961.1 | - | 1 | 2025 |