siRNA Details
| Gene Symbol: ARHGAP19-SLIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037909.1 | 15 | 1794 | |||
| GenBank mRNA | AY336750.1 | - | 15 | 1759 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK097919.1 | - | 1 | 2270 | Chr. 10 - 97153045 - 97155433 [-] on Build GRCh38 | |

ARHGAP19-SLIT1
Genomic Map