siRNA Details
| Gene Symbol: CSNK1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005256337.4 | XP_005256394.1 | 9 | 1989 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSNK1D| Gene Symbol: CSNK1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005256337.4 | XP_005256394.1 | 9 | 1989 | ||