siRNA Details
| Gene Symbol: LOC101928936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110867.1 | 3 | 1784 | |||
| GenBank mRNA | AK131305.1 | - | 3 | 1784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928936| Gene Symbol: LOC101928936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110867.1 | 3 | 1784 | |||
| GenBank mRNA | AK131305.1 | - | 3 | 1784 | ||