siRNA Details
| Gene Symbol: LOC101928936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110867.1 | 2 | 205 | |||
| GenBank mRNA | AK131305.1 | - | 2 | 205 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA721939.1 | - | 2 | 205 | Chr. 6 - 88529337 - 88539368 [-] on Build GRCh38 | |

LOC101928936
Genomic Map