siRNA Details
| Gene Symbol: ARID3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005259513.4 | XP_005259570.1 | 9 | 5300 | ||
| XM_005259514.4 | XP_005259571.1 | 9 | 4983 | |||
| XM_017026445.1 | XP_016881934.1 | 9 | 5009 | |||
| XM_017026446.1 | XP_016881935.1 | 7 | 4052 | |||
| XM_017026447.1 | XP_016881936.1 | 7 | 3892 | |||
| GenBank mRNA | BC010544.2 | - | 1 | 432 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN368274.1 | - | 1 | 50 | Chr. 19 - 974788 - 975114 [+] on Build GRCh38 | |

ARID3A
Genomic Map