siRNA Details
Gene Symbol: LINC00656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034149.1 | 4 | 768 | |||
GenBank mRNA | AF258576.1 | - | 4 | 753 | ||
BC117361.1 | - | 1 | 95 | 40125912 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034149.1 | 4 | 768 | |||
GenBank mRNA | AF258576.1 | - | 4 | 753 | ||
BC117361.1 | - | 1 | 95 | 40125912 |