siRNA Details
| Gene Symbol: LINC00656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034149.1 | 4 | 1018 | |||
| GenBank mRNA | AF258576.1 | - | 4 | 1005 | ||
| BC117361.1 | - | 1 | 345 | 40125912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00656| Gene Symbol: LINC00656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034149.1 | 4 | 1018 | |||
| GenBank mRNA | AF258576.1 | - | 4 | 1005 | ||
| BC117361.1 | - | 1 | 345 | 40125912 | ||