siRNA Details
| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006712677.3 | XP_006712740.1 | 7 | 4748 | ||
| GenBank mRNA | AK055724.1 | - | 1 | 1402 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPE| Gene Symbol: RPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006712677.3 | XP_006712740.1 | 7 | 4748 | ||
| GenBank mRNA | AK055724.1 | - | 1 | 1402 | ||