siRNA Details
| Gene Symbol: LOC105376545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC026354.1 | - | 3 | 1929 | ||
| Gene Symbol: ZBED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126323.1 | - | 5 | 1456 | ||

LOC105376545
Genomic Map