siRNA Details
| Gene Symbol: LINC01039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126390.1 | 4 | 3792 | |||
| GenBank mRNA | BC041830.1 | - | 5 | 3459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01039| Gene Symbol: LINC01039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126390.1 | 4 | 3792 | |||
| GenBank mRNA | BC041830.1 | - | 5 | 3459 | ||