siRNA Details
| Gene Symbol: PTPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005258124.3 | XP_005258181.1 | 10 | 3772 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK074657.1 | - | 1 | 61 | Chr. 18 - 12790117 - 12792062 [-] on Build GRCh38 | |

PTPN2
Genomic Map