siRNA Details
| Gene Symbol: NFYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK301215.1 | - | 5 | 1173 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK055329.1 | - | 1 | 1849 | Chr. 1 - 40765293 - 40767975 [+] on Build GRCh38 | |

NFYC
Genomic Map