siRNA Details
| Gene Symbol: PRR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534662.1 | XP_011532964.1 | 1 | 272 | ||
| XM_017009896.1 | XP_016865385.1 | 1 | 272 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125417.1 | - | 1 | 272 | Chr. 5 - 177448052 - 177452141 [+] on Build GRCh38 | |

PRR7
Genomic Map