siRNA Details
| Gene Symbol: PRR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174102.1 | NP_001167573.1 | 1 | 47 | ||
| XM_011534662.1 | XP_011532964.1 | 1 | 320 | |||
| XM_017009896.1 | XP_016865385.1 | 1 | 320 | |||
| GenBank mRNA | BC021240.1 | - | 1 | 38 | 3603954 | |
| BE277571.1 | - | 1 | 79 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125417.1 | - | 1 | 320 | Chr. 5 - 177448052 - 177452141 [+] on Build GRCh38 | |

PRR7
Genomic Map