siRNA Details
| Gene Symbol: RNASEH1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038429.1 | 2 | 784 | |||
| NR_038430.1 | 2 | 827 | ||||
| GenBank mRNA | AK023018.1 | - | 2 | 666 | ||
| BC019594.1 | - | 2 | 287 | |||
| BC040057.1 | - | 2 | 678 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ221780.1 | - | 2 | 676 | Chr. 2 - 3558494 - 3559676 [+] on Build GRCh38 | |

RNASEH1-AS1
Genomic Map