siRNA Details
| Gene Symbol: SPIRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130680.1 | - | 1 | 1140 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC035173.1 | - | 1 | 1938 | Chr. 16 - 89816769 - 89818757 [+] on Build GRCh38 | |

SPIRE2
Genomic Map