siRNA Details
| Gene Symbol: SPIRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011523379.1 | XP_011521681.1 | 1 | 543 | ||
| XM_011523381.1 | XP_011521683.1 | 1 | 543 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC035173.1 | - | 1 | 1255 | Chr. 16 - 89816769 - 89818757 [+] on Build GRCh38 | |

SPIRE2
Genomic Map