siRNA Details
| Gene Symbol: GBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006718047.1 | XP_006718110.1 | 9 | 1015 | ||
| XM_006718048.1 | XP_006718111.1 | 9 | 1015 | |||
| XM_006718049.1 | XP_006718112.1 | 9 | 1015 | |||
| XM_006718050.1 | XP_006718113.1 | 9 | 1015 | |||
| XM_011540312.1 | XP_011538614.1 | 9 | 1015 | |||
| XM_011540313.1 | XP_011538615.1 | 9 | 1015 | |||
| XM_017016862.1 | XP_016872351.1 | 9 | 1015 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023761.1 | - | 1 | 670 | Chr. 10 - 102353782 - 102356766 [+] on Build GRCh38 | |

GBF1
Genomic Map