siRNA Details
| Gene Symbol: LOC101928782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110836.1 | 3 | 574 | |||
| GenBank mRNA | AK098603.1 | - | 3 | 574 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415393.1 | - | 3 | 574 | Chr. 7 - 131910220 - 131948953 [+] on Build GRCh38 | |

LOC101928782
Genomic Map