siRNA Details
| Gene Symbol: DLEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AJ412056.1 | - | LIT1884 BCMS |
5 | 750 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AJ412005.3 | - | 1 | 13 | Chr. 13 - 50458002 - 50458113 [+] on Build GRCh38 | |

DLEU1
Genomic Map