siRNA Details
| Gene Symbol: LINC01095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038331.1 | 6 | 1131 | |||
| GenBank mRNA | AK057233.1 | - | 5 | 1666 | ||
| BC033400.1 | - | 6 | 1124 | |||
| DB525674.1 | - | 1 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01095| Gene Symbol: LINC01095 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038331.1 | 6 | 1131 | |||
| GenBank mRNA | AK057233.1 | - | 5 | 1666 | ||
| BC033400.1 | - | 6 | 1124 | |||
| DB525674.1 | - | 1 | 359 | |||